Learn More
Medchemexpress LLC Hki-357 | 848133-17-5 | MFCD26792555 | 99.2% | 574.05 g·mol⁻¹ | C31H29ClFN5O3 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10344325MG
Encompass_Preferred
HKI-357 is a small-molecule, irreversible dual inhibitor of EGFR and ERBB2 used for research applications. It shows low-nanomolar potency (IC50 EGFR 34 nM; ERBB2 33 nM) and suppresses EGFR autophosphorylation at Y1068 and downstream AKT and MAPK signaling. Intended for research use only.
  • Irreversible dual inhibition of EGFR and ERBB2 at low-nanomolar potency.
  • Suppresses EGFR autophosphorylation (Y1068) and downstream AKT and MAPK signaling.
  • Reported IC50s of 34 nM (EGFR) and 33 nM (ERBB2) for biochemical potency.
  • High purity suitable for in vitro research and biochemical assays.
  • Well-defined molecular formula and molecular weight facilitate analysis.
Catalog No. 50-002-44621