Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HKOCl-4 (BXY2142) | 2031170-85-9 | 95.1% | 1 MG
SDP

Supplier:  Medchemexpress LLC HY1300271MG

Encompass_Preferred

HKOCl-4 (BXY2142) is a rhodol-based yellow fluorescent probe designed for the detection of hypochlorous acid, offering excellent sensitivity and selectivity. It possesses a longer absorption wavelength and improved pH stability compared to fluorescein-based probes. This probe is intended for research use only.

  • Detection of hypochlorous acid
  • Excellent sensitivity and selectivity
  • Longer absorption wavelength
  • Improved pH stability
  • Excitation wavelength: 530 nm
  • Emission wavelength: 557 nm
  • Ultra-fast response to HOCl (reaches plateau within 2 min)
  • More than 20-fold increase in fluorescence intensity over other analytes
  • Non-toxic in living cells at concentrations up to 50 μM in RAW264.7 macrophages

Catalog No. 50-003-40571


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.