Learn More
Medchemexpress LLC Hksox-1r (5/6-mixture) | 98.38% | 100 MG
Supplier: Medchemexpress LLC HY130017100MG
HKSOX-1r (5/6-mixture) is a fluorescent probe utilized for the imaging and detection of endogenous superoxide in live cells and in vivo.
- Exhibits excellent selectivity towards superoxide anion radical.
- Demonstrates high sensitivity towards superoxide anion radical.