Learn More
Medchemexpress LLC HLA-DPB1 Human His 50ug
SDP
Supplier:  Medchemexpress LLC HYP7169650UG
Encompass_Preferred
Recombinant fragment of the human HLA-DPB1 (class II DP beta 1 chain) supplied as an N-terminal His-tagged, lyophilized protein for research use. The product corresponds to UniProt accession P04440 (residues R30-R223), has an approximate molecular weight of 26.8 kDa, and is >90% pure by reducing SDS-PAGE. Formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0, it is intended for biochemical and immunological applications and should be stored and handled according to the datasheet.
Catalog No. 50-001-95813