Learn More
Medchemexpress LLC HY-16667 5mg Medchemexpress, HLM006474 CAS:353519-63-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY166675MG
Encompass_Preferred
Medchemexpress, HY-16667 5mg HLM006474 CAS:353519-63-8 HLM006474 is a pan E2F inhibitor, which inhibits E2F4 DNA-binding with an IC50 of 29.8 µM in A375 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1899