Learn More
Sino Biological Human coronavirus (HCoV-NL63) Nucleocapsid Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40641T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human coronavirus (HCoV-NL63) Nucleoprotein / NP ( Catalog#40641-V07E; YP_003771.1; Met1-His377). Coronavirus Nucleocapsid specific IgG was purified by Human coronavirus (HCoV-NL63) Nucleoprotein / NP affinity chromatography.
Catalog No. NC2055730