Learn More
Abcam Human Cathepsin S ELISA Kit (CTSS), 1 x 96 Tests
Supplier:  Abcam AB1554271X96TESTS
Encompass_Preferred
Human Cathepsin S ELISA Kit (CTSS) is a Sandwich (quantitative) ELISA kit for the measurement of Human Cathepsin S (CTSS) in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2007718