Learn More
Abcam Human IgG ELISA Kit, Fluorescent, 1 x 96 Tests
Supplier:  Abcam AB2293901X96TESTS
Encompass_Preferred
Human IgG ELISA Kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human IgG with a sensitivity of 0.03 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1917791