Learn More
Neuromics, Inc Human Retinal Pigment Epithelial Cells, 500,000 Cells are initiated by dissecting retinal pigment tissue. The cells have a minimum average population doubling capacity >8.
Supplier:  Neuromics, Inc RPE001
Encompass
HRPEs are initiated by dissecting retinal pigment tissue and digestion with collagenase. HRPEs are then separated and purified before cryoperservation. Cells are provided at passage 3. HRPEs growth medium (contains 10% serum and growth supplements, Alpha-33) is recommended for cell culture and these cells have a minimum average population doubling capacity > 8 when cultured following the detailed protocol.
Catalog No. NC2066367