Learn More
Biocare Medical, LLC HMB45+MART-1+TYROSINASE 6ML
SDP
Supplier:  Biocare Medical, LLC PM165AA
Encompass_Preferred
HMB45 + MART-1 + Tyrosinase is a first-order pan melanoma screener. HMB45 labels the majority of melanomas. MART-1 is specific to melanocytic lesions. Tyrosinase labels a high percentage of desmoplastic melanomas.
Catalog No. 50-830-83