Learn More
Medchemexpress LLC Hydroxymethylbilane synthase (HMBS) protein | >95.0% | 10UG

Supplier: Medchemexpress LLC HYP70269Y10UG
Recombinant human hydroxymethylbilane synthase (HMBS), provided as a sterile, 0.22 μm filtered solution expressed in mammalian cells. The material is formulated with stabilizers and cryoprotectants to preserve activity and support biochemical assays, antibody validation, and enzymatic studies; it is supplied at high purity and concentration for reliable analytical use.
- Human sequence expressed in HEK293 cells.
- Includes C-terminal His tag for detection and purification.
- Purity greater than 95.0% by reducing SDS-PAGE.
- Supplied as a filtered solution in buffered formulation with cryoprotectants.
- Concentration provided at 1.44 mg/mL for flexible use and dilution.
- Available in small sizes suitable for analytical and control applications.
- Stable when stored at -80°C; avoid repeated freeze-thaw.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.