Learn More
Genetex Inc HMGA2 Monoclonal Antibody [HMV314] HistoMAX™ (GTX639938) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639938
Encompass_Preferred
The HMGA2 antibody [HMV314] HistoMAX™ (GTX639938) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human bronchial mucosa tissue using GTX639938 HMGA2 antibody [HMV314] HistoMAX. Strong nuclear HMGA2 staining of most respiratory epithelial cells.
Catalog No. NC3669315