Learn More
Genetex Inc HMGB1 Monoclonal Antibody [HMV317] HistoMAX™ (GTX639941) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639941
Encompass_Preferred
The HMGB1 antibody [HMV317] HistoMAX™ (GTX639941) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human esophageal squamous cell carcinoma (ESCC) tissue using GTX639941 HMGB1 antibody [HMV317] HistoMAX. Squamous cell carcinoma with strong HMGB1 staining of tumor cells.
Catalog No. NC3669319