Learn More
Labelmaster Inc Standard Hazcom (HMIS) Label, 2x2'', Paper
SDP
Supplier:  Labelmaster Inc HMIS0002
Encompass_Preferred
Our HMIS labels serve as a primary way to comply with OSHA's Hazard Communication Standard and have an easy-to-understand rating system to inform employees about the potential hazards of chemicals they might encounter in their workplace. Fill in these 2 x 2-in., high-gloss, thermal-imprintable, paper right to know labels with the Chemical Identity; the Health, Flammability, Physical Hazard ratings and the Personal Protection letter code for chemicals you use regularly.
Catalog No. NC1562676