Learn More
Medchemexpress LLC HMMNI-d3 (Hydroxy Dimetridazole-d3) | 1015855-78-3 | 160.15 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008216S10MG
Encompass_Preferred
HMMNI-d3 (Hydroxy Dimetridazole-d3) is a deuterium labeled form of HMMNI, which is a hydroxy metabolite of Dimetridazol. Dimetridazol is a nitroimidazole class agent effective against protozoan infections. This compound can be utilized as a tracer or as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled HMMNI
  • Hydroxy metabolite of Dimetridazol
  • Combats protozoan infections
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
Catalog No. 50-004-48783