Learn More
Medchemexpress LLC HMMNI (Hydroxy Dimetridazole) | 936-05-0 | 157.13 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00821610G
Encompass_Preferred
HMMNI (Hydroxy Dimetridazole) is a hydroxy metabolite of Dimetridazole, a nitroimidazole class drug. It is used to combat protozoan infections and exhibits antiprotozoal activity.
  • Effective against protozoan infections
  • Inhibits Trichomonas foetus with an ED50 of 1.6 μg/mL
  • Off-white to yellow solid appearance
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-48789