Learn More
Medchemexpress LLC HMMNI (Hydroxy Dimetridazole) | 936-05-0 | 157.13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW008216100MG
Encompass_Preferred
HMMNI (Hydroxy Dimetridazole) is a hydroxy metabolite of Dimetridazole, a nitroimidazole class drug. It is used to combat protozoan infections and is intended for research use only. This product is provided as a solid.
  • Used to combat protozoan infections
  • Intended for research use only
  • Off-white to yellow appearance
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-46072