Learn More
Medchemexpress LLC HMMNI (hydroxy dimetridazole) | 936-05-0 | 157.13 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00821650G
Encompass_Preferred
HMMNI (Hydroxy dimetridazole) is a hydroxy metabolite of Dimetridazole, a nitroimidazole class drug that combats protozoan infections. It demonstrates antiprotozoal activity.
  • Hydroxy metabolite
  • Combats protozoan infections
  • Exhibits antiprotozoal activity
  • Inhibits *Trichomonas foetus* with an ED50 of 1.6 μg/mL
Catalog No. 50-004-46189