Learn More
Medchemexpress LLC HY-12045 5mg Medchemexpress, HMN-214 CAS:173529-46-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY120455MG
Encompass_Preferred
Medchemexpress, HY-12045 5mg HMN-214 CAS:173529-46-9 HMN-214, an orally bioavailable prodrug of HMN-176, is an inhibitor of polo-like kinase-1 (plk1), with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0941