Learn More
Innovative Research Inc Hmn ATP5PB Prtn Lysate 20ug
Supplier:  Innovative Research Inc IHUATP5PBPLLY20UG
Encompass_Preferred
Human ATP5PB Protein Lysate 20ug - Human ATP5PB Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles.
Catalog No. 50-274-4356