Learn More
STEMCELL Technologies Human Peripheral Blood CD19+ B Cells, Frozen, Size: 1 x 10^7 cells
SDP
Supplier:  STEMCELL Technologies 70033
Encompass
Primary human CD19+ B cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using positive immunomagnetic separation techniques. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. NC1917571
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov