Learn More
Innovative Research Inc Hmn PAI1 Rcmb WT Act 500ug
Supplier:  Innovative Research Inc IHUPAI1ARWT500UG
Encompass_Preferred
Human PAI-1 Recombinant Wild Type Active 500 ug - Human PAI-1 Recombinant Wild Type Active from Innovative Research has been recombinantly produced in E. coli and purified by affinity chromatography. This is a Frozen liquid buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6 and a purity of >95% as determined by SDS-PAGE. More Details Species Human Target PAI-1 Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source E. coli Storage Conditions -70C Additional Information Gentle purification conditions create an active fraction that is greater than 99% pure and 98% active. This product can be used as a control in applications like ELISA and Western Blot. It can be used in a wide range of in vitro applications, including blocking/inhibition, fibrinolysis, plasminogen activation, receptor binding, in hypertension studies, for protein-protein interactions, and more.
Catalog No. 50-274-4288