Learn More
STEMCELL Technologies Human Peripheral Blood CD4+CD45RA+ T Cells, Frozen, Size: 5 x 10^6 cells
SDP
Supplier:  STEMCELL Technologies 70029
Encompass
Primary human CD4+CD45RA+ T cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. PB was collected using acid-citrate-dextrose solution A (ACDA) as the anticoagulant. CD4 and CD45RA are co-expressed on naïve CD4+ T cells.Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. NC0662293
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov