Learn More
Innovative Research Inc Hmn Secretory IgA Pur Lyo 5mg
Supplier:  Innovative Research Inc IHUSIGALY5MG
Encompass_Preferred
Human Secretory IgA Purified Lyophilized 5mg - Human Secretory IgA Purified Lyophilized from Innovative Research has been purified from human colostrum. This product is offered in a lyophilized powder format, buffered in 0.02M Na Phosphate; 0.15M NaCl; pH 7.4, with a purity of >95% by SDS-PAGE analysis. Secretory Immunoglobulin A is the primary immunoglobulin on most mucosal surfaces. SigA is a polypetide complex made up of two IgA monomers, a connecting J chain, and the secretory component. Located on mucosal surfaces, SigA provides protection by preventing invasion of pathogens. No reaction by IEP with antiserum to human Albumin, IgD, IgE, IgG, IgM and Lactoferrin. >95percent pure by SDS-PAGE. Add deionized water to original volume, aliquot and freeze unused portion.
Catalog No. 50-274-4256