Learn More
Innovative Research Inc Hmn U2OS Whl Cell Lys 500ug
Supplier:  Innovative Research Inc IHUU2OSTLW500UG
Encompass_Preferred
Human U2OS Whole Cell Lysate 500ug - Human U2OS Whole Cell Lysate from Innovative Research is provided as a Liquid. Store at -70C or below. If not using the entire volume immediately, store single-use aliquots to avoid repeated freeze/thaw cycles.
Catalog No. 50-274-4243