Learn More
Medchemexpress LLC Hmnni (hydroxy dimetridazole) | 936-05-0 | 157.13 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW008216500MG
Encompass_Preferred
HMMNI (Hydroxy dimetridazole) is a hydroxy metabolite of Dimetridazole. Dimetridazole is a nitroimidazole class drug that combats protozoan infections.
  • Exhibits antiprotozoal activity.
  • Inhibits Trichomonas foetus with an ED50 of 1.6 μg/mL.
Catalog No. 50-004-47430