Learn More
Sino Biological Human metapneumovirus (HMPV) Fusion glycoprotein (His Tag) (MALS-verified)
Supplier:  Sino Biological 40993V08H1100
Encompass_Preferred
A DNA sequence encoding the Human metapneumovirus (HMPV)(strain CAN97-83) Fusion glycoprotein (Q6WB98) with the preFusion design was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3303