Learn More
Lonza Walkersville HMVEC-dAd – Human Dermal Microvascular Endothelial Cells – Adult
Supplier:  Lonza Walkersville CC2543
Encompass
Human Dermal Microvascular Endothelial Cells include cells isolated from small vessels within skin tissue from either adults or neonatal foreskins.Cells are cultured to be ≥90% pure and specifically enriched for dermal LECs and BECs. Dermal microvascular endothelial cells, from either neonates or adults (HMVEC-D) are a mixed population of both blood (HMVEC-DBl) and lymphatic (HMVEC-DLy) endothelial cells.Benefits:Ready-To-Use No tissue dissection required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batch Content:1 x Cryopreserved Ampule with ≥500,000 cells/vial
Catalog No. NC9507316