Learn More
Neuromics, Inc Human Retinal Microvascular Endothelial Cells, 500,000 Cells were isolated from normal human retinal tissue. Cells have an average additional population doubling level >16 when cultured.
Supplier:  Neuromics, Inc HEC09
Encompass
HRMECs were initiated by elutriation from dissociated normal human retinal tissue. Passage 3 cells are shipped frozen. ENDO-Growth medium is recommended for culture. Cells have an average additional population doubling levels >16 when cultured.Characterization of the cells: Cytoplasmic VWF / Factor VIII: >95% positive by immunofluorescence, Cytoplasmic uptake of Di-I-Ac-LDL: >95% positive by immunofluorescence, and Cytoplasmic PECAM1 >95% positive by immunofluorescenceHRMECs are negative for HIV-1, HBV, HCV, and mycoplasma.
Catalog No. NC2071948