Learn More
Medchemexpress LLC HY-111798 5mg Medchemexpress, HO-1-IN-1 CAS:1093058-52-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1117985MG
Encompass_Preferred
Medchemexpress, HY-111798 5mg HO-1-IN-1 CAS:1093058-52-6 HO-1-IN-1 (Compound 2) is a heme oxygenase 1 ( HO-1 ) inhibitor with an IC 50 of 250 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2396