Learn More
Medchemexpress LLC HY-111798A 10mg Medchemexpress, HO-1-IN-1 hydrochloride CAS:1092851-70-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111798A10MG
Encompass_Preferred
Medchemexpress, HY-111798A 10mg HO-1-IN-1 HCl CAS:1092851-70-1 HO-1-IN-1 hydrochloride (Compound 2) is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 250 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8970