Learn More
Medchemexpress LLC HO-2/HMOX2 Human H 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP79126100UG
Encompass_Preferred
Through its catalytic activity HO-2/HMOX2 protein promotes the oxidative cleavage of heme at the -methylene bridge carbon thereby releasing carbon monoxide (CO) This process simultaneously produces biliverdin IXalpha and releases the central heme iron chelate in the form of ferrous iron HO-2/HMOX2 Protein Human (His) is the recombinant human-derived HO-2/HMOX2 protein expressed by E coli with C-6 His labeled tag
Catalog No. 50-001-94979