Learn More
Medchemexpress LLC HO-PEG16-OH | 6812-36-8 | 98.0% | C32H66O17 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY141235500MG
Encompass_Preferred
HO-PEG16-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • Facilitates protein degradation through the ubiquitin-proteasome system.
  • Composed of two ligands connected by a linker.
  • Targets specific proteins for selective degradation.
Catalog No. 50-003-53200