Learn More
Medchemexpress LLC Ho-peg16-oh | 6812-36-8 | MFCD09910395 | 98.0% | C32H66O17 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141235
Encompass_Preferred
HO-PEG16-OH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs are heterobifunctional nanomolecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • Appearance: Solid, white to off-white
  • Functions as a PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52750