Learn More
Promocell GmbH PromoCell HOB-c 500,000 Human Osteoblasts, cryopreserved
Supplier:  Promocell GmbH C12720
Encompass
Primary Human Osteoblasts (HOB) are isolated from femoral trabecular bone tissue from the knee or hip joint region (lot specific source information is available on request). They stain positive for alkaline phosphatase and can be mineralized using PromoCell Mineralization Medium.Osteoblasts are of mesenchymal origin and are mainly responsible for bone formation and remodeling. In vivo, they produce a collagen type I rich extracellular matrix called the osteoid.During their development, osteoblasts undergo a maturation process called osteogenesis. When they become encased in the bone matrix, they stop proliferating and become osteocytes. Our HOB are now also available from HLA-typed donors.
Catalog No. 50-306-059