Learn More
Apexbio Technology LLC HOBt (anhydrous), 250g Cas No: 2592-95-2 MFCD: MFCD00005805
SDP
Supplier:  Apexbio Technology LLC A7025250000
Encompass_Preferred
Hydroxy benzotriazole, abbreviated HOBt, is anorganic compoundthat is a derivative of benzotriazole. HOBt, as a commercial product, is a white crystalline powder contains some water (~11.7% wt as the HOBt monohydrate crystal). There are other sizes available. Please inqury us for quote.
Catalog No. NC1572002