Learn More
AdipoGen Hoechst 33342
SDP
Supplier:  AdipoGen CDXB0030G005
Encompass_Preferred
Chemical. CAS 23491-52-3. Formula C27H28N6O . 3HCl . xH2O. MW 561.93. Synthetic. Useful for staining DNA, chromosomes and nuclei. May be used for fluorescence microscopy or flow cytometry. Suitable for vital DNA staining of a variety of cell types and as a probe of membrane permeability in mammalian cells.
Catalog No. 50-256-1701