Learn More
Cayman Chemical Hoechst 33342hydroCLOrID 100mg
SDP
Supplier:  Cayman Chemical 15547100 MG
Encompass_Preferred
A cell-permeable, fluorescent probe for DNA that is useful as a marker of nuclei for cell cycle studies in combination with BrdU and to distinguish nuclear morphology in apoptotic cells
Catalog No. 50-297-3344