Learn More
Medchemexpress LLC Hoipin-8 | 2519537-70-1 | 98.9% | 100 MG

Supplier: Medchemexpress LLC HY122882100MG
HOIPIN-8 is a small-molecule inhibitor of the linear ubiquitin chain assembly complex (LUBAC) that selectively inhibits HOIP and suppresses NF-κB signaling; reported IC50 ≈ 11 nM. Supplied as an off-white to light yellow solid sodium salt for in vitro research.
- Potent LUBAC (HOIP) inhibitor with reported IC50 ≈ 11 nM.
- Sodium salt; molecular weight 456.38 g/mol; formula C23H15F2N4NaO3.
- High purity (98.9%) solid suitable for biochemical and cell-based studies.
- Soluble in DMSO at approximately 50 mg/mL; ultrasonic assistance recommended.
- Store sealed, away from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.