Learn More
Selleck Chemical LLC HOIPIN-8-E1798-5MG
SDP
Supplier:  Selleck Chemical LLC E17985MG
Encompass_Preferred
HOIPIN-8 is a potent inhibitor of linear ubiquitin chain assembly complex (LUBAC) and NF- B signaling with an IC50 value of 11 nM It serves as a powerful tool for investigating the physiological and cellular functions of LUBAC
Catalog No. 50-313-9121