Learn More
Enterprise Technology Solutions Dynarex Blood Collection Tube Holder
SDP
Supplier:  Enterprise Technology Solutions DRX6933
Encompass_Preferred
The Dynarex Blood Collection Tube Holders have a slim, easy to handle design, enhancing the users control and flexibility. These tube holders are made of a translucent plastic for easy viewing of blood collection while providing a secure connection to collection tubes, needles, and sets when performing a direct draw into a blood collection tube. The tube holders are available with a Luer Lock or Luer Slip connection.
Catalog No. 50-240-7078