Learn More
JDM LAB SOLUTIONS LLC Holder for chromatography column
Supplier:  JDM LAB SOLUTIONS LLC 01139
Encompass
The module allows effortless attachment of chromatography columns. The bracket adjusts to the column circumference via a snap-in mechanism to provide a secure fit. For lengths greater than 15 cm of the suspended equipment, the use of two modules arranged one above the other is recommended to ensure the stability of the superstructure.
Catalog No. 50-255-8567