Learn More
Med Vet International Webster Needle Holder 5-in Smooth Jaws Standard Pattern Von Klaus German Surgical Steel
Supplier:  Med Vet International VK1101014
Encompass_Preferred
Webster Needle Holder 5-in Smooth Jaws Standard Pattern Von Klaus German Surgical Steel
Catalog No. 50-307-2765