Learn More
Med Vet International Olsen-Hegar Needle Holder & Scissors, 4.5in, Tungsten Carbide, Serrated, German Grade, Von Klaus
Supplier:  Med Vet International VK132159
Encompass_Preferred
4.25in. Olsen-Hegar Needle holders is a German grade surgical instrument and tungsten carbide. They are serrated and are generally used with sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2278