Learn More
Med Vet International Olsen-Hegar Needle Holder, 5.5 in, Von Klaüs German Surgical Steel
Supplier:  Med Vet International VK1106014
Encompass_Preferred
Olsen-Hegar needle holders is a German surgical steel instrument. They feature serrated jaws, integrated scissors and are generally used with sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2282