Learn More
MSC M.K. MORSE Bi-Metal Hole Saw, 1-1/8 in, M42, 1 ea
Supplier:  MSC 32552655
Encompass
This bi-metal hole saw is engineered for general purpose cutting across a wide range of materials, including metals, wood, drywall, and composites. It features a 1-1/8 in diameter and a toothed cutting edge, representing an optimized solution for various applications.
  • Patent pending tooth set design optimized to remove material faster
  • Premium M42 high speed steel cutting edge with 8% cobalt for extended life
  • Solid cap reduces runout and vibration during operation
  • Heavy duty 0.05 in side wall for greater stability
  • New side slot increases leverage for faster, easier slug removal
  • Exterior red coating reduces side wall friction for efficient cutting
  • Variable pitch pattern with 5/6 teeth per inch
  • Cutting depth of 1.9370 in
  • Applicable for a wide range of materials including various metals, woods, plastics, and composites
Catalog No. 50-192-0196