Learn More
MSC M.K. MORSE Hole Saw, 3-1/4 in Dia, Bi-Metal, 1 ea.
Supplier:  MSC 32550659
Encompass
This bi-metal hole saw features a toothed cutting edge, designed for efficient cutting through a variety of materials. It is suitable for applications involving wood, stainless steel, aluminum, machinable metal, plastic, and nail-embedded wood.
  • Saw material: Bi-metal
  • Cutting depth: 1-15/16 in
  • Cutting edge style: Toothed
  • Material application: Wood, stainless steel, aluminum, machinable metal, plastic, nail-embedded wood
  • Pitch pattern: Variable
  • Teeth per inch: 18
  • Material grade: M42
  • Arbor not included
Catalog No. 50-192-0200