Learn More
Electron Microscopy Sciences C-Flat™ Holey Carbon Grid Gold 2.0 µm Hole 2.0 µm Space 200 Mesh, 50/pack
SDP
Supplier:  Electron Microscopy Sciences CF222C50AU
Encompass_Preferred
C-Flat™ Holey Carbon Grid Gold 2.0 µm Hole 2.0 µm Space 200 Mesh.C-flat™ is the premier holey carbon grid for cryo-transmission electron microscopy.C-flat™ is an ultra-flat, holey carbon-coated TEM support grid for transmission electron microscopy (TEM). Unlike competing holey carbon films, C-flat™ is manufactured without plastics, so it is clean upon arrival and the user has no residue to contend with.
Catalog No. NC2605661