Learn More
Strem, An Ascensus Company CAS# 14483-18-2. 5g. Holmium(III) nitrate hydrate (99.9%-Ho) (REO). MFCD00149737
SDP
Supplier:  Strem, An Ascensus Company 9367125G
Encompass
CAS# 14483-18-2. 5g. Holmium(III) nitrate hydrate (99.9%-Ho) (REO). MFCD00149737. Molecular Weight: 350.95. Molecular Formula: Ho(NO3)3∙XH2O. Color/form: orange xtl. Strem# 93-6712. http://www.strem.com/catalog/v/93-6712/
Catalog No. 50-901-16139