Learn More
Aviva Systems Biology Corp HOM-TES-103 Antibody - C-terminal region (ARP55421_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55421_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HOM-TES-103. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HOM-TES-103. Alternate names: DKFZp586I2223, FLJ20703, HOM-TES-103, IFFO, MGC117359ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-035